missing translation for 'onlineSavingsMsg'
Learn More

Cyclopamine, MedChemExpress

Product Code. p-200097590
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16463287

Brand: Medchem Express HY17024

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cyclopamine is a Hedgehog (Hh) pathway antagonist with an IC50 of 46 nM in the Hh cell assay. Cyclopamine is also a selective Smo inhibitor.

Cyclopamine is a Hedgehog (Hh) pathway antagonist with an IC50 of 46 nM in the Hh cell assay. Cyclopamine is also a selective Smo inhibitor.

Specifications

Chemical Name or Material Cyclopamine
CAS 4449-51-8
Color White
Purity Grade Notes Research
Molecular Formula C27H41NO2
Quantity 50 mg
Synonym 11-Deoxojervine
Solubility Information Ethanol : 20 mg/mL (48.59 mM; Need ultrasonic) ∣DMSO : 10 mg/mL (24.29 mM; ultrasonic and warming and heat to 80°C)
SMILES CC1=C2[C@@]([C@@]3([H])[C@]([C@@]4(C(C[C@@H](O)CC4)=CC3)C)([H])C2)([H])CC[C@@]15[C@@H]([C@@]6([H])[C@](C[C@H](C)CN6)([H])O5)C
Molecular Weight (g/mol) 411.62
Formula Weight 411.62
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.