missing translation for 'onlineSavingsMsg'
Learn More

Cyclopamine, MedChemExpress

Product Code. 16463287
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463287 50 mg 50mg
16437236 100 mg 100mg
16417278 10 mg 10mg
16409797 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16463287 Supplier Medchem Express Supplier No. HY17024

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cyclopamine is a Hedgehog (Hh) pathway antagonist with an IC50 of 46 nM in the Hh cell assay. Cyclopamine is also a selective Smo inhibitor.

Cyclopamine is a Hedgehog (Hh) pathway antagonist with an IC50 of 46 nM in the Hh cell assay. Cyclopamine is also a selective Smo inhibitor.

Specifications

Chemical Name or Material Cyclopamine
CAS 4449-51-8
Color White
Purity Grade Notes Research
Molecular Formula C27H41NO2
Quantity 50 mg
Synonym 11-Deoxojervine
Solubility Information Ethanol : 20 mg/mL (48.59 mM; Need ultrasonic) ∣DMSO : 10 mg/mL (24.29 mM; ultrasonic and warming and heat to 80°C)
SMILES CC1=C2[C@@]([C@@]3([H])[C@]([C@@]4(C(C[C@@H](O)CC4)=CC3)C)([H])C2)([H])CC[C@@]15[C@@H]([C@@]6([H])[C@](C[C@H](C)CN6)([H])O5)C
Molecular Weight (g/mol) 411.62
Formula Weight 411.62
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.