missing translation for 'onlineSavingsMsg'
Learn More

Cyclopamine, 99%

Product Code. 15494109
Click to view available options
Quantity:
25 mg
50 mg
Unit Size:
25mg
50mg
This item is not returnable. View return policy

Product Code. 15494109

Brand: Thermo Scientific Alfa Aesar J61528.MF

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4449-51-8 | C27H41NO2 | 411.63 g/mol

Cyclopamine, is used as a hedgehog [Hh] signaling pathway and Smo inhibitor. It depletes stem-like cancer cells in glioblastoma and blocks tumor engraftment. Can also be used to induce differentiation of human embryonic stem cells (hESCs) into hormone-expressing endocrine cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopamine, is used as a hedgehog [Hh] signaling pathway and Smo inhibitor. It depletes stem-like cancer cells in glioblastoma and blocks tumor engraftment. Can also be used to induce differentiation of human embryonic stem cells (hESCs) into hormone-expressing endocrine cells.

Solubility
Soluble in ethanol (5 mg/ml warm), DMSO (4 mg/ml), DMF (10 mg/ml), methylene chloride, chloroform, methanol (7 mg/ml), and benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4449-51-8
Molecular Formula C27H41NO2
Molecular Weight (g/mol) 411.63
MDL Number MFCD01735266
InChI Key QASFUMOKHFSJGL-LAFRSMQTSA-N
Synonym cyclopamine, 3b,23b-17,23-epoxy-11-deoxoveratraman-3-ol
PubChem CID 122130428
SMILES C[C@@H]1[C@@H]2NC[C@@H](C)C[C@H]2O[C@]11CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC2=C1C

Specifications

Melting Point 234°C to 237°C
Color White
Quantity 50 mg
Solubility Information Soluble in ethanol (5mg/ml warm),DMSO (4mg/ml),DMF (10mg/ml),methylene chloride,chloroform,methanol (7mg/ml),and benzene.
IUPAC Name (3S,3'R,3'aS,6'S,6aS,6bS,7'aR,9R,11aS,11bR)-3',6',10,11b-tetramethyl-1,2,3,3'a,4,4',5',6,6',6a,6b,7,7',7'a,8,11,11a,11b-octadecahydro-3'H-spiro[cyclohexa[a]fluorene-9,2'-furo[3,2-b]pyridin]-3-ol
Formula Weight 411.62
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material Cyclopamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.