missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Cyclopamine
Description
- Cyclopamine is a steroid alkaloid that inhibits the Hedgehog pathway at the point of the pathway activator Smoothened
- Cyclopamine binds to the heptahelical bundle of Smoothened, a G protein-coupled receptor, and prevents it from signaling further downstream (Chen et al.)
- Reduces proliferation of rat neural progenitor cells and mouse neurospheres (Lai et al.; Palma and Ruiz i Altaba)
- Reduces proliferation of mouse mammospheres (Liu et al.)
- Promotes differentiation of pancreatic cells from human embryonic stem cells (D'Amour et al.)
- Inhibits the growth of human and mouse medulloblastoma cells, and human glioblastoma cells (Bar et al.), Berman et al.)
Specifications
Specifications
| CAS | 4449-51-8 |
| Synonym | 11-Deoxojervine; Jervine |
| Quantity | 1 mg |
| Molecular Formula | C27H41NO2 |
| Purity | ≥95% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?