missing translation for 'onlineSavingsMsg'
Learn More

Cyclooctylamine, 97%

Product Code. 10646914 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10646914 100 g Glass Bottle 100g
10775941 5 g Glass Bottle 5g
10172420 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10646914 Supplier Thermo Scientific Acros Supplier No. 111461000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5452-37-9 | C8H17N | 127.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5452-37-9
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.23
MDL Number MFCD00001748
InChI Key HSOHBWMXECKEKV-UHFFFAOYSA-N
Synonym cyclooctylamine, aminocyclooctane, cyclooctyl amine, acmc-1ap2c, hsohbwmxeckekv-uhfffaoysa, cyclooctylamine, purum gc
PubChem CID 2903
IUPAC Name cyclooctanamine
SMILES C1CCCC(CCC1)N

Specifications

Melting Point -48°C
Color Colorless to Yellow
Density 0.9200g/mL
Boiling Point 190°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula C8H15NH2
Refractive Index 1.4800 to 1.482
Quantity 100 g
Beilstein 12, IV, 128
Specific Gravity 0.92
Solubility Information Solubility in water: 10.2g/L (20°C)
Viscosity 5 mPa.s (20°C)
Formula Weight 127.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Cyclooctylamine
Show More Show Less
compliance-icons
Product Identifier
  • Cyclooctylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.