missing translation for 'onlineSavingsMsg'
Learn More

cis-Cyclooctene, 95%, stabilized

Product Code. 10255530 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10255530 1 L Glass bottle 1 L
10011800 250 mL Glass bottle 250mL
11974431 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10255530 Supplier Thermo Scientific Acros Supplier No. 154860010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 931-87-3 | C8H14 | 110.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 931-87-3
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number MFCD00001753
InChI Key URYYVOIYTNXXBN-UPHRSURJSA-N
Synonym cis-cyclooctene, z-cyclooctene, cyclooctene, z, cyclooctene, 1z, cyclooct-1-ene, e-cyclooctene, 1-cyclooctene #, 1z-cyclooctene, 7z-cyclooctene, ar,e-cyclooctene
PubChem CID 638079
SMILES C1CCC\C=C/CC1

Specifications

Melting Point -16.0°C
Color Colorless to Brown
Density 0.8400g/mL
Boiling Point 145.0°C to 146.0°C
Flash Point 25°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula C8H14
Refractive Index 1.4680 to 1.4710
Quantity 1 L
Specific Gravity 0.84
Solubility Information Solubility in water: <0.1g/L (20°C)
IUPAC Name cyclooctene
Stabilizer 100 to 200ppm irganox 1076 FD
Viscosity 1.89 mPa.s (20°C)
Formula Weight 110.2
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material cis-Cyclooctene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.