missing translation for 'onlineSavingsMsg'
Learn More

Cyclooctatetraene, TRC

Product Code. 30349084
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349084 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30349084 Supplier Toronto Research Chemicals Supplier No. TRCC9897205G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclooctatetraene
CAS 629-20-9
Molecular Formula C8 H8
Quantity 5 g
Synonym 1,3,5,7-Cyclooctatetraene, Cyclooctatetraene, NSC 5093, [8]Annulene
InChI Formula InChI=1S/C8H8/c1-2-4-6-8-7-5-3-1/h1-8H/b2-1-,3-1-,4-2-,5-3-,6-4-,7-5-,8-6-,8-7-
SMILES C1\C=C/C=C\C=C/C=1
IUPAC Name cyclooctatetraene
Molecular Weight (g/mol) 104.149
Formula Weight 104.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.