missing translation for 'onlineSavingsMsg'
Learn More

Cyclooctanone, TRC

Product Code. 30365991
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365991 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30365991 Supplier Toronto Research Chemicals Supplier No. TRCC988345250G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclooctanone
CAS 502-49-8
Molecular Formula C8H14O
Quantity 250 g
Synonym NSC 9475
InChI Formula InChI=1S/C8H14O/c9-8-6-4-2-1-3-5-7-8/h1-7H2
SMILES O=C1CCCCCCC1
IUPAC Name cyclooctanone
Molecular Weight (g/mol) 126.2
Formula Weight 126.1045
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.