missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclooctanol, 97%
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
Cyclooctanol is a key starting material used for the preparation of cylooctanone by using a chromic acid reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cyclooctanol is a key starting material used for the preparation of cylooctanone by using a chromic acid reagent.
Solubility
Miscible with acetone.
Notes
Contains 75+% cyclopentane; remainder consists of C{5} and C{6} hydrocarbon.Incompatible with strong oxidizing agents, acid chloride, and acid anhydrides.
Cyclooctanol is a key starting material used for the preparation of cylooctanone by using a chromic acid reagent.
Solubility
Miscible with acetone.
Notes
Contains 75+% cyclopentane; remainder consists of C{5} and C{6} hydrocarbon.Incompatible with strong oxidizing agents, acid chloride, and acid anhydrides.
Specifications
Specifikationer
| Melting Point | 14°C to 15°C |
| Density | 0.97 |
| Boiling Point | 97°C to 98°C (11 mmHg) |
| Flash Point | 86°C (186°F) |
| Refractive Index | 1.486 |
| Quantity | 100 g |
| Beilstein | 1848166 |
| Solubility Information | Miscible with acetone. |
| Formula Weight | 128.22 |
| Percent Purity | 97% |
| Visa mer |
RUO – Research Use Only
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering