Learn More
Specac™ Cyclone™ C5 Long Pathlength Gas Cell GKFV
Product Code. 15230278
Encompass_Preferred
Click to view available options
Volume (Metric):
1.33 L
Product Code. 15230278
Brand: Specac™ GS24105GKFV
This is a heatable long pathlength gas cell with glass body, KBr windows, fixed pathlength and Viton O-rings. The cell has a volume of 1.33 litres.
Specifications
Description Fixed pathlength and Viton O-rings
Volume (Metric) 1.33 L