Learn More
Specac™ Cyclone™ C5 Long Pathlength Gas Cell GKAV
Product Code. 15220278
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
1.33 L
Product Code. Volume (Metric)
15220278 1.33 L
1 options
Product Code. 15220278 Supplier Specac™ Supplier No. GS24105GKAV
Only null left
Add to basket
Add to basket
This is a heatable long pathlength gas cell with glass body, KBr windows, adjustable pathlength and Viton O-rings. The pathlength is adjustable from 1m - 8m in steps of 1m and the cell has a volume of 1.33 litres.
Specifications
Description Adjustable pathlength and Viton O-rings
Volume (Metric) 1.33 L