Learn More
Specac™ Cyclone™ C2 Long Pathlength Gas Cell MZFV
Product Code. 15290268
Encompass_Preferred
Click to view available options
Volume (Metric):
0.19 L
Product Code. 15290268
Brand: Specac™ GS24102MZFV
This is a heatable long pathlength gas cell with metal body, ZnSe windows, fixed pathlength and Viton O-rings. The cell has a volume of 0.19 litres.
Specifications
Description Fixed pathlength and Viton O-rings
Volume (Metric) 0.19 L