Learn More
Specac™ Cyclone™ C2 Long Pathlength Gas Cell MZAV
Product Code. 15280268
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
0.19 L
Product Code. Volume (Metric)
15280268 0.19 L
1 options
Product Code. 15280268 Supplier Specac™ Supplier No. GS24102MZAV
Add to basket
Add to basket
This is a heatable long pathlength gas cell with metal body, ZnSe windows, adjustable pathlength and Viton O-rings. The pathlength is adjustable from 0.5-2.5m in 0.5m steps and has a volume of 0.19 litres.
Specifications
Description Adjustable pathlength and Viton O-rings
Volume (Metric) 0.19 L