Learn More
Specac™ Cyclone™ C2 Long Pathlength Gas Cell GKFV
Product Code. 15230268
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
0.19 L
Product Code. Volume (Metric)
15230268 0.19 L
1 options
Product Code. 15230268 Supplier Specac™ Supplier No. GS24102GKFV
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This is a heatable long pathlength gas cell with glass body, KBr windows, fixed pathlength and Viton O-rings. The cell has a volume of 0.19 litres.
Specifications
Description Fixed pathlength and Viton O-rings
Volume (Metric) 0.19 L