Learn More
Specac™ Cyclone™ C2 Long Pathlength Gas Cell GKAV
Product Code. 15220268
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
0.19 L
Product Code. Volume (Metric)
15220268 0.19 L
1 options
Product Code. 15220268 Supplier Specac™ Supplier No. GS24102GKAV
Only null left
Add to basket
Add to basket
This is a heatable long pathlength gas cell with glass body, KBr windows, adjustable pathlength and Viton O-rings. The pathlength is adjustable from 0.5-2.5m in steps of 0.5m with a volume of 0.19 litres.
Specifications
Description Adjustable pathlength and Viton O-rings
Volume (Metric) 0.19 L