Learn More
Specac™ Cyclone™ C10 Long Pathlength Gas Cell MZFV
Product Code. 15290288
Encompass_Preferred
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
2.61 L
Product Code. Volume (Metric)
15290288 2.61 L
1 missing translation for 'options'
Product Code. 15290288 missing translation for 'mfr' Specac™ missing translation for 'supplierNo' GS24110MZFV
Estimated Shipment: 16-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This is a heatable long pathlength gas cell with metal body, ZnSe windows, fixed pathlength and Viton O-rings. The cell has a volume of 2.61 litres.
Specifications
Volume (Metric) 2.61 L