Learn More
Specac™ Cyclone™ C10 Long Pathlength Gas Cell MCFV
Product Code. 15270288
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
2.61 L
Product Code. Volume (Metric)
15270288 2.61 L
1 options
Product Code. 15270288 Supplier Specac™ Supplier No. GS24110MCFV
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This is a heatable long pathlength gas cell with metal body, CaF2 windows, fixed pathlength and Viton O-rings. The cell has a volume of 2.61 litres.
Specifications
Description Fixed pathlength and Viton O-rings
Volume (Metric) 2.61 L