Learn More
Specac™ Cyclone™ C10 Long Pathlength Gas Cell GZAV
Product Code. 15240288
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
2.61 L
Product Code. Volume (Metric)
15240288 2.61 L
1 options
Product Code. 15240288 Supplier Specac™ Supplier No. GS24110GZAV
Only null left
Add to basket
Add to basket
This is a heatable long pathlength gas cell with glass body, ZnSe windows, adjustable pathlength and Viton O-rings. The pathlength is adjustable from 2.1m - 10.6m in steps of 1.06m and the cell has a volume of 2.61 litres.
Specifications
Description Adjustable pathlength and Viton O-rings
Volume (Metric) 2.61 L