Learn More
Specac™ Cyclone™ C10 Long Pathlength Gas Cell GKAV
Product Code. 15220288
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
2.61 L
Product Code. Volume (Metric)
15220288 2.61 L
1 options
Product Code. 15220288 Supplier Specac™ Supplier No. GS24110GKAV
Estimated Shipment: 14-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This is a heatable long pathlength gas cell with glass body, KBr windows, adjustable pathlength and Viton O-rings. The pathlength is adjustable from 2.1m - 10.6m in steps of 1.06m and the cell has a volume of 2.61 litres.
Specifications
Description Adjustable pathlength and Viton O-rings
Volume (Metric) 2.61 L