missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyltriphenylphosphonium bromide, 98+%

Product Code. 11422477
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11422477 5 g 5g
11432477 25 g 25g
11442477 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11422477 Supplier Thermo Scientific Alfa Aesar Supplier No. A14298.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7333-51-9 | C24H26BrP | 425.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7333-51-9
Molecular Formula C24H26BrP
Molecular Weight (g/mol) 425.35
MDL Number MFCD00031667
InChI Key QRAKRDZMIONMRZ-UHFFFAOYSA-M
Synonym cyclohexyltriphenylphosphonium bromide, cyclohexyl triphenylphosphonium bromide, cyclohexyltriphenylphosphanium bromide, cyclohexyl triphenyl phosphanium bromide, acmc-20alcd, cyclohexyltriphenylphosphoniumbromide, cyclohexyl triphenyl phosphonium bromide
PubChem CID 2724861
SMILES [Br-].C1CCC(CC1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 265°C to 272°C
Quantity 5 g
Beilstein 3581012
Sensitivity Hygroscopic
IUPAC Name cyclohexyltriphenylphosphanium bromide
Formula Weight 425.36
Percent Purity ≥98%
Chemical Name or Material Cyclohexyltriphenylphosphonium bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.