missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylmethyldichlorosilane, 97+%, Thermo Scientific Chemicals

Product Code. 10412790 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
This item is not returnable. View return policy

Product Code. 10412790

Brand: Thermo Scientific Acros 432220500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5578-42-7
Molecular Formula C7H14Cl2Si
Molecular Weight (g/mol) 197.18
InChI Key YUYHCACQLHNZLS-UHFFFAOYSA-N
Synonym methylcyclohexyldichlorosilane, silane, dichlorocyclohexylmethyl, dichloro cyclohexyl methyl silane, cyclohexylmethyldichlorosilane, cyclohexane, dichloromethylsilyl, methyl cyclohexyldichlorosilane, cyclohexyl methyl dichlorosilane, cyclohexyl-dichloro-methylsilane, cyclohexyl-methyl-dichlorosilane, dichloro cyclohexyl methylsilane
PubChem CID 79685
IUPAC Name dichloro-cyclohexyl-methylsilane
SMILES C[Si](C1CCCCC1)(Cl)Cl

Specifications

Color Colorless to Brown
Density 1.0950g/mL
Boiling Point 202.0°C
Flash Point 66°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula C6H11Si(CH3)Cl2
Refractive Index 1.4710 to 1.4730 (20°C, 589nm)
Quantity 50 mL
Specific Gravity 1.095
Solubility Information Solubility in water: reacts
Formula Weight 197.18
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material Cyclohexylmethyldichlorosilane
Show More Show Less
compliance-icons
Product Identifier
  • Cyclohexylmethyldichlorosilane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.