missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylmethyl bromide, 96%

Product Code. 10681195 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10681195 25 g Glass bottle 25g
10253832 5 g Glass bottle 5g
10220753 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10681195 Supplier Thermo Scientific Acros Supplier No. 111350250
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2550-36-9 | C7H13Br | 177.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2550-36-9
Molecular Formula C7H13Br
Molecular Weight (g/mol) 177.09
MDL Number MFCD00001509
InChI Key UUWSLBWDFJMSFP-UHFFFAOYSA-N
Synonym bromomethyl cyclohexane, cyclohexylmethyl bromide, cyclohexane, bromomethyl, bromocyclohexylmethane, cyclohexylmethylbromide, cyclohexylmethybromide, bromomethyl-cyclohexane, pubchem3103, 1-bromomethylcyclohexane
PubChem CID 137636
SMILES BrCC1CCCCC1

Specifications

Color Yellow
Density 1.2800g/mL
Boiling Point 76°C to 77°C (26.0 mmHg)
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula C6H11CH2Br
Refractive Index 1.4910 to 1.493
Quantity 25 g
Beilstein 05, IV, 100
Specific Gravity 1.28
IUPAC Name bromomethylcyclohexane
Formula Weight 177.08
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Cyclohexylmethyl bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.