missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylmagnesium chloride, 1M in MeTHF

Product Code. 15488047
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488047 500 mL 500mL
15478047 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15488047 Supplier Thermo Scientific Alfa Aesar Supplier No. H51165.AP
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 931-51-1 | C6H11ClMg | 142.91 g/mol

Cyclohexylmagnesium chloride is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF). It is used as a reagent for the introduction of the cyclohexyl group; metalation of CH-acidic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexylmagnesium chloride is used as a Grignard reagent in greener solvent, 2-methyltetrahydrofuran (2-MeTHF). It is used as a reagent for the introduction of the cyclohexyl group; metalation of CH-acidic compounds.

Solubility
It reacts violently with water.

Notes
Air & moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 931-51-1
Molecular Formula C6H11ClMg
Molecular Weight (g/mol) 142.91
MDL Number MFCD00003816
InChI Key WMJMABVHDMRMJA-UHFFFAOYSA-M
Synonym cyclohexylmagnesium chloride, magnesium, chlorocyclohexyl, magnesium,chlorocyclohexyl, cyclohexyl magnesium chloride, chloro cyclohexyl magnesium, chlorocyclohexylmagnesium, cyclohexylmagnesiumchloride, unii-oag686rdsu, cyclohexylmagnesium chloride solution, 2.0 m in diethyl ether, cyclohexylmagnesium cloride
PubChem CID 13605
IUPAC Name magnesium;cyclohexane;chloride
SMILES Cl[Mg]C1CCCCC1

Specifications

Quantity 500 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Solubility Information It reacts violently with water.
Formula Weight 142.91
Concentration or Composition (by Analyte or Components) 1M in MeTHF
Chemical Name or Material Cyclohexylmagnesium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.