missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyldiphenylphosphine, 98%

Product Code. 11344237
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344237 100 g 100g
15451467 1 g 1g
11334237 5 g 5g
11324237 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11344237 Supplier Thermo Scientific Alfa Aesar Supplier No. 036708.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6372-42-5 | C18H21P | 268.34 g/mol

Cyclohexyldiphenylphosphine is used as ligand in coupling reaction. It is also used as a medical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6372-42-5
Molecular Formula C18H21P
Molecular Weight (g/mol) 268.34
MDL Number MFCD00046360
InChI Key ZXKWUYWWVSKKQZ-UHFFFAOYSA-N
Synonym cyclohexyldiphenylphosphine, phosphine, cyclohexyldiphenyl, cyclohexyl diphenyl phosphine, diphenylcyclohexylphosphine, diphenylphosphinocyclohexane, cyclohexyl-diphenyl-phosphane, pubchem6451, acmc-1ba4g, cyclohexyl diphenyl phosphane
PubChem CID 80756
SMILES C1CCC(CC1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 58°C to 62°C
Flash Point >110°C (230°F)
Quantity 100 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
IUPAC Name cyclohexyl(diphenyl)phosphane
Formula Weight 268.34
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Cyclohexyldiphenylphosphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.