missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylbenzene, 97+%

Product Code. 11427797
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11427797 500 g 500g
11417797 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11427797 Supplier Thermo Scientific Alfa Aesar Supplier No. A16320.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 827-52-1 | C12H16 | 160.26 g/mol

Cyclohexylbenzene is used as a high boiling point solvent. It finds application as a penetrant used in plastics, painting, and adhesives. It acts as an additive for lithium-ion battery electrolyte to enhance the battery safety. It is used to prepare intermediates such as phenol and cyclohexanone. Further, it is used in the preparation of liquid crystal display (LCD) material intermediate 4-ethyl cyclohexyl benzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexylbenzene is used as a high boiling point solvent. It finds application as a penetrant used in plastics, painting, and adhesives. It acts as an additive for lithium-ion battery electrolyte to enhance the battery safety. It is used to prepare intermediates such as phenol and cyclohexanone. Further, it is used in the preparation of liquid crystal display (LCD) material intermediate 4-ethyl cyclohexyl benzoic acid.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 827-52-1
Molecular Formula C12H16
Molecular Weight (g/mol) 160.26
MDL Number MFCD00001451
InChI Key IGARGHRYKHJQSM-UHFFFAOYSA-N
Synonym phenylcyclohexane, benzene, cyclohexyl, cyclohexane, phenyl, cyclohexyl-benzene, cyclohexyl benzene, 4-cyclohexylbenzene, santosol 360, phenyl cyclohexane, 1,1'-biphenyl, 1,2,3,4,5,6-hexahydro, wln: l6tj ar
PubChem CID 13229
IUPAC Name cyclohexylbenzene
SMILES C1CCC(CC1)C1=CC=CC=C1

Specifications

Melting Point 5°C to 6°C
Density 0.945
Boiling Point 239°C to 240°C
Flash Point 98°C (208°F)
Odor Mild
Refractive Index 1.526
Quantity 500 g
UN Number UN3082
Beilstein 1906803
Solubility Information Immiscible with water.
Formula Weight 160.26
Percent Purity ≥97%
Chemical Name or Material Cyclohexylbenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.