missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylamine, 98+%

Product Code. 11314539
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11314539 100 mL 100mL
11402548 500 mL 500mL
11412548 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11314539 Supplier Thermo Scientific Alfa Aesar Supplier No. A15851.AE
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 108-91-8 | C6H13N | 99.177 g/mol

Cyclohexylamine is a useful as an intermediate in organic synthesis. It acts as a precursor to prepare pendant-armed dialdehydes, bromohexine HCl and sulfenamide-based reagents. It is widely used in the production of vulcanization accelerators like N-cyclohexyl-2-benzothiazole sulfonamide (CBS) and artificial sweeteners (cyclamates). As a corrosion inhibitor, it is used in the treatment of boiler water and prevents corrosion of metal surfaces that arise due to dissolved carbon dioxide. It is involved as a solvent in the synthesis of an inorganic-organic hybrid semiconductor, ZnS/CHA (CHA = cyclohexylamine) nanocomposites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexylamine is a useful as an intermediate in organic synthesis. It acts as a precursor to prepare pendant-armed dialdehydes, bromohexine HCl and sulfenamide-based reagents. It is widely used in the production of vulcanization accelerators like N-cyclohexyl-2-benzothiazole sulfonamide (CBS) and artificial sweeteners (cyclamates). As a corrosion inhibitor, it is used in the treatment of boiler water and prevents corrosion of metal surfaces that arise due to dissolved carbon dioxide. It is involved as a solvent in the synthesis of an inorganic-organic hybrid semiconductor, ZnS/CHA (CHA = cyclohexylamine) nanocomposites.

Solubility
Miscible with water, ethanol, ethers, acetone, alcohol and ketones.

Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents, carbon dioxide, sodium hypochlorite, organic acids, mineral acids and peroxides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-91-8
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.177
MDL Number MFCD00001486
InChI Key PAFZNILMFXTMIY-UHFFFAOYSA-N
Synonym cyclohexylamine, aminocyclohexane, hexahydroaniline, hexahydrobenzenamine, aminohexahydrobenzene, cyclohexyl amine, 1-cyclohexylamine, 1-aminocyclohexane, aniline, hexahydro, benzenamine, hexahydro
PubChem CID 7965
ChEBI CHEBI:15773
IUPAC Name cyclohexanamine
SMILES C1CCC(CC1)N

Specifications

Melting Point -18°C
Density 0.868
Boiling Point 133°C to 134°C
Flash Point 27°C (81°F)
Refractive Index 1.4585
Quantity 100 mL
UN Number UN2357
Beilstein 471175
Sensitivity Air Sensitive
Merck Index 14,2729
Solubility Information Miscible with water,ethanol,ethers,acetone,alcohol and ketones.
Formula Weight 99.18
Percent Purity ≥98%
Chemical Name or Material Cyclohexylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.