missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylacetylene, 98%

Product Code. 15440208
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15440208 5 g 5g
15450208 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15440208 Supplier Thermo Scientific Alfa Aesar Supplier No. H53418.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 931-48-6 | C8H12 | 108.184 g/mol

Cyclohexylacetylene is used in the preparation of hydrido-vinylidene complexes of osmium, which finds vital intermediates in several homogeneous and heterogeneous catalytic reactions such as alkene olimerization, polymerization and Fischer-Tropsch synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexylacetylene is used in the preparation of hydrido-vinylidene complexes of osmium, which finds vital intermediates in several homogeneous and heterogeneous catalytic reactions such as alkene olimerization, polymerization and Fischer-Tropsch synthesis.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 931-48-6
Molecular Formula C8H12
Molecular Weight (g/mol) 108.184
MDL Number MFCD00001513
InChI Key SSDZYLQUYMOSAK-UHFFFAOYSA-N
Synonym cyclohexylacetylene, cyclohexylethyne, cyclohexane, ethynyl, cyclohexyl acetylene, ethynyl-cyclohexane, cyclohexane, ethynyl-6ci, 7ci, 8ci, 9ci
PubChem CID 70263
IUPAC Name ethynylcyclohexane
SMILES C#CC1CCCCC1

Specifications

Density 0.828
Boiling Point 130°C to 132°C
Flash Point 18°C (64°F)
Refractive Index 1.454
Quantity 5 g
UN Number UN3295
Beilstein 1815535
Solubility Information Immiscible with water.
Formula Weight 108.18
Percent Purity 98%
Chemical Name or Material Cyclohexylacetylene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.