missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylacetone, 95%

Product Code. 11470713
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470713 25 g 25g
11460713 5 g 5g
11480713 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470713 Supplier Thermo Scientific Alfa Aesar Supplier No. A12748.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 103-78-6 | C9H16O | 140.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 103-78-6
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.23
MDL Number MFCD00019396
InChI Key UHBZEAPZATVYKV-UHFFFAOYSA-N
Synonym cyclohexylacetone, 1-cyclohexylacetone, 2-propanone, 1-cyclohexyl, acetonylcyclohexane, cyclohexyl-2-propanone, 1-cyclohexyl-2-propanone, cyclohexylaceton, acmc-20alcn, 1-cyclohexylacetone #, 1-cyclohexyl-propan-2-one
PubChem CID 66899
IUPAC Name 1-cyclohexylpropan-2-one
SMILES CC(=O)CC1CCCCC1

Spécification

Density 0.905
Boiling Point 198°C to 200°C
Refractive Index 1.4525
Quantity 25 g
Beilstein 1209723
Formula Weight 140.23
Percent Purity 97%
Chemical Name or Material Cyclohexylacetone

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.