missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl ethyl ketone, 99%

Product Code. 11454337
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454337 5 g 5g
11464337 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454337 Supplier Thermo Scientific Alfa Aesar Supplier No. B20409.06
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1123-86-0 | C9H16O | 140.226 g/mol

Cyclohexyl ethyl ketone is used as a intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexyl ethyl ketone is used as a intermediate in organic synthesis.

Solubility
Soluble in organic solvents.

Notes
Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1123-86-0
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.226
MDL Number MFCD00060814
InChI Key AMHOPTNGSNYSBL-UHFFFAOYSA-N
Synonym ethyl cyclohexyl ketone, cyclohexylethylketone, 1-propanone, 1-cyclohexyl, cyclohexyl ethyl ketone, ketone, cyclohexyl ethyl, 1-cyclohexyl-1-propanone, 1-cyclohexyl-propan-1-one, propanoylcyclohexane, 1-methylacetylcyclohexan, acmc-1c9ub
PubChem CID 70748
IUPAC Name 1-cyclohexylpropan-1-one
SMILES CCC(=O)C1CCCCC1

Specifications

Density 0.91
Quantity 5 g
Solubility Information Soluble in organic solvents.
Formula Weight 140.23
Percent Purity 99%
Chemical Name or Material Cyclohexyl ethyl ketone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.