missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl acrylate, 98+%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals

Product Code. p-7046049
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11377796

Brand: Thermo Scientific Alfa Aesar L12925.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3066-71-5
Molecular Formula C9H14O2
Molecular Weight (g/mol) 154.21
MDL Number MFCD00046349
InChI Key KBLWLMPSVYBVDK-UHFFFAOYSA-N
Synonym cyclohexyl acrylate, 2-propenoic acid, cyclohexyl ester, sartomer sr 220, acrylic acid, cyclohexyl ester, acrylic acid cyclohexyl ester, cyclohexylacrylate, acrylic acid cyclohexyl, acmc-209slu, wln: l6tj aov1u1, 4-06-00-00038 beilstein handbook reference
PubChem CID 18298
IUPAC Name cyclohexyl prop-2-enoate
SMILES C=CC(=O)OC1CCCCC1

Specifications

Boiling Point 182°C to 184°C
Flash Point 68°C (154°F)
Quantity 5 g
UN Number UN3082
Beilstein 1937528
Formula Weight 154.21
Percent Purity ≥98%
Chemical Name or Material Cyclohexyl acrylate, stabilized with 100ppm 4-methoxyphenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.