missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl acrylate, 98+%, stab. with 100ppm 4-methoxyphenol, Thermo Scientific Chemicals

Product Code. 11377796
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11377796 5 g 5g
11387796 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377796 Supplier Thermo Scientific Alfa Aesar Supplier No. L12925.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3066-71-5 | Molecular Formula: C9H14O2 | Molecular Weight: 154.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3066-71-5
Molecular Formula C9H14O2
Molecular Weight (g/mol) 154.21
MDL Number MFCD00046349
InChI Key KBLWLMPSVYBVDK-UHFFFAOYSA-N
Synonym cyclohexyl acrylate, 2-propenoic acid, cyclohexyl ester, sartomer sr 220, acrylic acid, cyclohexyl ester, acrylic acid cyclohexyl ester, cyclohexylacrylate, acrylic acid cyclohexyl, acmc-209slu, wln: l6tj aov1u1, 4-06-00-00038 beilstein handbook reference
PubChem CID 18298
IUPAC Name cyclohexyl prop-2-enoate
SMILES C=CC(=O)OC1CCCCC1

Specifications

Boiling Point 182°C to 184°C
Flash Point 68°C (154°F)
Quantity 5 g
UN Number UN3082
Beilstein 1937528
Formula Weight 154.21
Percent Purity ≥98%
Chemical Name or Material Cyclohexyl acrylate, stabilized with 100ppm 4-methoxyphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.