missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl 2-chloroacetate, 97%, Thermo Scientific™

£61.00 - £161.00

Chemical Identifiers

CAS 6975-91-3
Molecular Formula C8H13ClO2
Molecular Weight (g/mol) 176.64
MDL Number MFCD00084989
InChI Key IBFHBLOKQVCABG-UHFFFAOYSA-N
Synonym cyclohexyl chloroacetate, cyclohexyl chloroacetate #, 1-chloroacetyl oxy cyclohexane, chloroacetic acid cyclohexyl ester, chloroacetic acid, cyclohexyl ester, acetic acid, 2-chloro-, cyclohexyl ester
PubChem CID 228008
IUPAC Name cyclohexyl 2-chloroacetate
SMILES C1CCC(CC1)OC(=O)CCl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10426662
View Documents Promotion Details
Thermo Scientific Maybridge
KM03519DA
1 g Amber Glass Bottle
£61.00
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10436662
View Documents Promotion Details
Thermo Scientific Maybridge
KM03519EA
10 g Amber Glass Bottle
£161.00
10g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

6975-91-3
176.64
IBFHBLOKQVCABG-UHFFFAOYSA-N
228008
C1CCC(CC1)OC(=O)CCl
C8H13ClO2
MFCD00084989
cyclohexyl chloroacetate, cyclohexyl chloroacetate #, 1-chloroacetyl oxy cyclohexane, chloroacetic acid cyclohexyl ester, chloroacetic acid, cyclohexyl ester, acetic acid, 2-chloro-, cyclohexyl ester
cyclohexyl 2-chloroacetate

Specifications

6975-91-3
97%
ClCH2CO2C6H11
cyclohexyl chloroacetate, cyclohexyl chloroacetate #, 1-chloroacetyl oxy cyclohexane, chloroacetic acid cyclohexyl ester, chloroacetic acid, cyclohexyl ester, acetic acid, 2-chloro-, cyclohexyl ester
C1CCC(CC1)OC(=O)CCl
176.64
176.64
Cyclohexyl 2-chloroacetate
97.0
C8H13ClO2
MFCD00084989
IBFHBLOKQVCABG-UHFFFAOYSA-N
cyclohexyl 2-chloroacetate
228008
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications