missing translation for 'onlineSavingsMsg'
Learn More

Cycloheximide, TRC

Product Code. 30366239
Change view
Click to view available options
Quantity:
100 mg
1 g
2.5 g
Unit Size:
100mg
1g
2.50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30366239 2.5 g 2.50g
30360263 1 g 1g
30362943 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30366239 Supplier Toronto Research Chemicals Supplier No. TRCC9882702.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66-81-9
Molecular Formula C15 H23 N O4
Molecular Weight (g/mol) 281.35
Synonym 2,6-Piperidinedione, 4-[2-(3,5-dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]-, [1S-[1α(S*),3α,5β]]-, Glutarimide, 3-[2-(3,5-dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]- (7CI,8CI), Naramycin A (6CI), 4-[(2R)-2-[(1S,3S,5S)-3,5-Dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]-2,6-piperidinedione, (-)-Cycloheximide, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide, Acti-Aid, Acti-dione, Acti-dione BR, Acti-dione PM, Acti-dione TGF, Actidion, CHX, Cicloheximide, Cycloheximide, FT 3422-2, LDN 97549, NM-MCD 80, NSC 185, Naramycin, Naramycin Microcapsule D 80, Naramycin NM-MCU 80, TZA, U 4527, l-Cycloheximide, β-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide
IUPAC Name 4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione
SMILES C[C@H]1C[C@H](C)C(=O)[C@@H](C1)[C@H](O)CC2CC(=O)NC(=O)C2

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C15H23NO4/c1-8-3-9(2)15(20)11(4-8)12(17)5-10-6-13(18)16-14(19)7-10/h8-12,17H,3-7H2,1-2H3,(H,16,18,19)/t8-,9-,11-,12+/m0/s1
Formula Weight 281.1627
Chemical Name or Material Cycloheximide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.