missing translation for 'onlineSavingsMsg'
Learn More

Melford Cycloheximide

Product Code. 30018211
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018211 5 g 5g
30018209 1 g 1g
30018210 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30018211 Supplier Melford Supplier No. C810405.0
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cycloheximide is a fungal-derived compound used particularly in molecular biology, microbiology, and cell biology.

Cycloheximide is a fungal-derived compound used particularly in molecular biology, microbiology, and cell biology. It inhibits protein synthesis in eukaryotic cells by interfering with the translocation step during protein elongation on the ribosome. It binds to the 60S ribosomal subunit and prevents the movement of peptidyl-tRNA from the A-site to the P-site, inhibiting the synthesis of new polypeptide chains.

Widely used in research to study cellular processes, including apoptosis, autophagy, and the regulation of gene expression. Cycloheximide is often used in cell culture studies to block de novo protein synthesis, allowing researchers to study the stability and turnover of existing proteins. In microbiology research, it selectively inhibits the growth of fungi in culture media, making it a useful tool for isolating and identifying bacteria without interference from fungi. ​​​​​

Specifications

Chemical Name or Material Cycloheximide
CAS 66-81-9
Quantity 5 g
Molecular Formula C15H23NO4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.