missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexene, TRC

Product Code. 30352805
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
This item is not returnable. View return policy

Product Code. 30352805

Brand: Toronto Research Chemicals TRCC9875135ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclohexene
CAS 110-83-8
Purity Grade Notes GC
Molecular Formula C6H10
Quantity 5 mL
Synonym 1,2,3,4-Tetrahydrobenzene, Benzene tetrahydride, Benzene, tetrahydro-, Cyclohex-1-ene, NSC 24835, Tetrahydrobenzene
InChI Formula InChI=1S/C6H10/c1-2-4-6-5-3-1/h1-2H,3-6H2
SMILES C1CCC=CC1
IUPAC Name cyclohexene
Molecular Weight (g/mol) 82.14
Formula Weight 82.0783
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.