Learn More
Cyclohexene, 99% stab.
Description
Cyclohexene is used as a solvent. It is employed as a precursor to caprolactam, adipic acid, maleic acid, dicyclohexyladipate and cyclohexeneoxide. It is used as an intermediate for the production of cyclohexanol and cyclohexeneoxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Cyclohexene is used as a solvent. It is employed as a precursor to caprolactam, adipic acid, maleic acid, dicyclohexyladipate and cyclohexeneoxide. It is used as an intermediate for the production of cyclohexanol and cyclohexeneoxide.
Solubility
Immiscible with cold water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | -104°C |
| Density | 0.811 |
| Boiling Point | 83°C to 85°C |
| Flash Point | −20°C (−4°F) |
| Odor | Petrol-like |
| Refractive Index | 1.446 |
| Quantity | 500 mL |
| UN Number | UN2256 |
| Beilstein | 906737 |
| Merck Index | 14,2727 |
| Show More |
Safety and Handling
- Cyclohexene
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H304-May be fatal if swallowed and enters airways.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P331-Do NOT induce vomiting.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- MIXTURE LIST-Contain: Cyclohexene
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.