missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone, ACS, Greenfield

Product Code. 30286176
Change view
Click to view available options
Quantity:
4x4L Case
5 Gallon
Unit Size:
16 L
18.93 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30286176 4x4L Case 16 L
30286185 5 Gallon 18.93 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30286176 Supplier Pharmco Supplier No. 314000ACSCS4L
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 108-94-1

Specifications

Quantity 4x4L Case
Packaging 4x4L Case
Boiling Point 156°C
Grade Reagent
Chemical Name or Material Cyclohexanone, ACS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.