missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone, ACS, 99+%

Product Code. 11474990
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
Unit Size:
1 L
4 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474990 500 mL 500mL
11387628 1 L 1 L
11307638 4 x 1 L 4 L
11397628 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11474990 Supplier Thermo Scientific Alfa Aesar Supplier No. 033309.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-94-1 | C6H10O | 98.145 g/mol

Cyclohexanone is used as an industrial solvent and paint remover. It acts as a precursor to nylon 6,6 and nylon 6 and cyclohexanone oxime, which gives caprolactam on rearrangement. Further, it is used as a chemical reaction medium, adhesives, sealants and agricultural products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-94-1
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00001625
InChI Key JHIVVAPYMSGYDF-UHFFFAOYSA-N
Synonym ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon
PubChem CID 7967
ChEBI CHEBI:17854
IUPAC Name cyclohexanone
SMILES C1CCC(=O)CC1

Specifications

Melting Point °C to 26°C
Color Colorless
Appearance Colorless liquid
Density 0.947 g/cm3 at 20°C
Quantity 500 mL
Flash Point 47°C (116°F)
Physical Form Liquid
Refractive Index 1.4507
Assay Percent Range >99%
Vapor Pressure 4.5 hPa (3mm Hg) at 20°C
Beilstein 385735
Boiling Point 154°C to 156°C
Solubility Information Miscible with water and ethanol,acetone,ethyl ether and other organic solvents.
Formula Weight 98.15
Grade ACS
Chemical Name or Material Cyclohexanone, For ACS analysis
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.