missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone, 99+%

Product Code. 11467477
Change view
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467477 500 mL 500mL
11477477 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11467477 Supplier Thermo Scientific Alfa Aesar Supplier No. A15607.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-94-1 | C6H10O | 98.145 g/mol

Cyclohexanone is used in the preparation of synthetic polymer such as nylon 6,6 and nylon 6. It acts as a solvent in organic synthesis. Its derivatives are used for the synthesis of dyes, rubber chemicals, plasticizers and pharmaceuticals. Further, it is involved in the preparation of cyclohexanone oxime, which rearranges to give caprolactam in the presence of sulfuric acid catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanone is used in the preparation of synthetic polymer such as nylon 6,6 and nylon 6. It acts as a solvent in organic synthesis. Its derivatives are used for the synthesis of dyes, rubber chemicals, plasticizers and pharmaceuticals. Further, it is involved in the preparation of cyclohexanone oxime, which rearranges to give caprolactam in the presence of sulfuric acid catalyst.

Solubility
Miscible with diethyl ether, acetone and alcohol. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, red metals, lead, amines, sulfuric acid, nitric acid, strong acids, aliphatic amines, rubber, resins and alkalies.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-94-1
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00001625
InChI Key JHIVVAPYMSGYDF-UHFFFAOYSA-N
Synonym ketohexamethylene, pimelic ketone, sextone, cyclohexyl ketone, nadone, anone, anon, cyclohexanon, hytrol o, hexanon
PubChem CID 7967
ChEBI CHEBI:17854
IUPAC Name cyclohexanone
SMILES C1CCC(=O)CC1

Specifications

Melting Point °C to 26°C
Density 0.9478
Quantity 500 mL
Flash Point 47°C (116°F)
Physical Form Liquid
Refractive Index 1.4507
Assay Percent Range ≥99%
UN Number UN1915
Beilstein 385735
Merck Index 14,2726
Boiling Point 154°C to 156°C
Solubility Information Miscible with diethyl ether,acetone and alcohol. Slightly miscible with water.
Formula Weight 98.15
Percent Purity ≥99%
Chemical Name or Material Cyclohexanone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.