missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexanone, 99+%, ACS reagent
Sfoglia Tutto Thermo Scientific Chemicals Prodotti
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Dimensione della confezione:
1 L
2.5 L
250mL
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Melting Point | -47.0°C |
| Density | 0.9470g/mL |
| Boiling Point | 155.0°C |
| Quantity | 2.5 L |
| Flash Point | 43°C |
| Refractive Index | 1.4450 to 1.4560 |
| Specific Gravity | 0.947 |
| Linear Formula | C6H10(=O) |
| Water | 0.05% max. |
| Solubility Information | Solubility in water: 150g/L (10°C). Other solubilities: soluble in all common organic solvents |
| Vedi altri risultati |
Sicurezza e movimentazione
Identificativo prodotto
- Cyclohexanone
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto