missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexanone-2,2,6,6-d4, CDN
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Cyclohexanone-2,2,6,6-D4 |
| CAS | 1006-03-7 |
| Molecular Formula | C6 D4 H6 O |
| Quantity | 1 g |
| Synonym | Cyclohexanone-2,2,6,6-d4 (6 CI, 7 CI, 8 CI, 9 CI, ACI), 2,2,6,6-Tetradeuteriocyclohexanone, [2,2,6,6-d4]-Cyclohexanone, Cyclohexanone-2,2,6,6-D4, Cyclohexanone-d4 |
| InChI Formula | InChI=1 S/C6H10O/c7-6-4-2-1-3-5-6/h1-5H2/i4D2,5D2 |
| SMILES | [2 H]C1([2 H])CCCC([2 H])([2 H])C1=O |
| IUPAC Name | 2,2,6,6-tetradeuteriocyclohexan-1-one |
| Molecular Weight (g/mol) | 102.1677 |
| Formula Weight | 102.0983 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction