missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanol, TRC

Product Code. 30132751
Change view
Click to view available options
Quantity:
250 mL
500 mL
1 L
Unit Size:
1 L
250mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132751 1 L 1 L
30136875 250 mL 250mL
30011787 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132751 Supplier Toronto Research Chemicals Supplier No. TRCC9879601L

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-93-0
Molecular Formula C6 H12 O
Molecular Weight (g/mol) 100.16
Synonym 1-Cyclohexanol, Adronal, Adronol, Anol, Cyclohexyl alcohol, Hexahydrophenol, Hexalin, Hexalin (alcohol), Hydroxycyclohexane, NSC 403656, NSC 54711, Naxol, Phenol, hexahydro-
IUPAC Name cyclohexanol
SMILES OC1CCCCC1

Specifications

Quantity 1 L
InChI Formula InChI=1S/C6H12O/c7-6-4-2-1-3-5-6/h6-7H,1-5H2
Formula Weight 100.0888
Chemical Name or Material Cyclohexanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.