missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanemethylamine, 98%

Product Code. 11322546
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322546 25 g 25g
11312546 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322546 Supplier Thermo Scientific Alfa Aesar Supplier No. L06501.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3218-02-8 | C7H15N | 113.20 g/mol

Cyclohexanemethylamine was used in the synthesis of Schiff?s base ligand, 4-methyl-2,6-bis(cyclohexylmethyliminomethyl)-phenol, which forms hexanuclear zinc(II) complexes. It was used during the synthesis of 1-acyl-4 cycloalkyl/arylsemicarbazides and 1-acyl-5-benzyloxy/hydroxyl carbamoylcarbazides from the nonsteroidal anti-inflammatory drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanemethylamine was used in the synthesis of Schiff′s base ligand, 4-methyl-2,6-bis(cyclohexylmethyliminomethyl)-phenol, which forms hexanuclear zinc(II) complexes. It was used during the synthesis of 1-acyl-4 cycloalkyl/arylsemicarbazides and 1-acyl-5-benzyloxy/hydroxyl carbamoylcarbazides from the nonsteroidal anti-inflammatory drugs.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3218-02-8
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.20
MDL Number MFCD00001520
InChI Key AVKNGPAMCBSNSO-UHFFFAOYSA-N
Synonym cyclohexanemethylamine, aminomethylcyclohexane, aminomethyl cyclohexane, cyclohexanemethanamine, 1-cyclohexylmethanamine, hexahydrobenzylamine, 1-cyclohexylmethylamine, 4-aminomethylcyclohexane, c-cyclohexyl-methylamine, chembl1049
PubChem CID 76688
IUPAC Name cyclohexylmethanamine
SMILES C1CCC(CC1)CN

Specifications

Melting Point -8°C
Density 0.871
Boiling Point 161°C to 163°C
Flash Point 43°C (109°F)
Refractive Index 1.462
Quantity 25 g
UN Number UN2734
Beilstein 635751
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 113.2
Percent Purity 98%
Chemical Name or Material Cyclohexanemethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.