missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanemethanol, 99%

Product Code. 11342556
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342556 100 g 100g
11332556 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342556 Supplier Thermo Scientific Alfa Aesar Supplier No. L06538.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-49-2 | C7H14O | 114.188 g/mol

Cyclohexanemethanol used as solvents, fuels . It is also used in pharmaceuticals, plasticizers, surfactants, lubricants, ore floatation agents, hydraulic fluids, and detergents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanemethanol used as solvents, fuels . It is also used in pharmaceuticals, plasticizers, surfactants, lubricants, ore floatation agents, hydraulic fluids, and detergents.

Solubility
Soluble in Ether, Alcohols.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents, Acid chlorides, Acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-49-2
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.188
MDL Number MFCD00001510
InChI Key VSSAZBXXNIABDN-UHFFFAOYSA-N
Synonym cyclohexanemethanol, cyclohexylcarbinol, hexahydrobenzyl alcohol, cyclohexanecarbinol, methanol, cyclohexyl, cyclohexylmethyl alcohol, benzyl alcohol, hexahydro, usaf do-49, hydroxymethylcyclohexane, hydroxymethyl cyclohexane
PubChem CID 7507
IUPAC Name cyclohexylmethanol
SMILES C1CCC(CC1)CO

Specifications

Density 0.926
Boiling Point 180°C to 181°C
Flash Point 71°C (159°F)
Refractive Index 1.464
Quantity 100 g
Beilstein 773712
Merck Index 14,2731
Solubility Information Soluble in Ether,Alcohols.
Formula Weight 114.19
Percent Purity 99%
Chemical Name or Material Cyclohexanemethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.