missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanecarbonyl chloride, 97+%

Product Code. 11448677
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448677 100 g 100g
11438677 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11448677 Supplier Thermo Scientific Alfa Aesar Supplier No. A19824.22
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2719-27-9 | C7H11ClO | 146.61 g/mol

Cyclohexanecarbonyl chloride is used in the preparation of five thiourea derivative ligands having anti-bacterial and anti-yeast activity, (N-(diethylcarbamothioyl)cyclohexanecarboxamide, N-(di-n-propylcarbamothioyl)cyclohexanecarboxamide, di-n-butylcarbamothioyl)cyclohexanecarboxamide, N-(diphenylcarbamothioyl)cyclohexanecarboxamide, N-(morpholine-4-carbonothioyl)cyclohexanecarboxamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexanecarbonyl chloride is used in the preparation of five thiourea derivative ligands having anti-bacterial and anti-yeast activity, (N-(diethylcarbamothioyl)cyclohexanecarboxamide, N-(di-n-propylcarbamothioyl)cyclohexanecarboxamide, di-n-butylcarbamothioyl)cyclohexanecarboxamide, N-(diphenylcarbamothioyl)cyclohexanecarboxamide, N-(morpholine-4-carbonothioyl)cyclohexanecarboxamide.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2719-27-9
Molecular Formula C7H11ClO
Molecular Weight (g/mol) 146.61
MDL Number MFCD00001456
InChI Key RVOJTCZRIKWHDX-UHFFFAOYSA-N
Synonym hexahydrobenzoyl chloride, chlorocarbonylcyclohexane, cyclohexanoyl chloride, cyclohexylcarbonyl chloride, cyclohexoyl chloride, cyclohexancarbonylchlorid, acmc-209gvv, hexahydrobenzoyl-chloride, cyclohexanecarbonylchloride, cyclohexancarbonyl chloride
PubChem CID 75938
IUPAC Name cyclohexanecarbonyl chloride
SMILES ClC(=O)C1CCCCC1

Specifications

Density 1.088
Boiling Point 183°C to 185°C
Flash Point 66°C (150°F)
Refractive Index 1.47
Quantity 100 g
UN Number UN3265
Beilstein 742163
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 146.62
Percent Purity ≥97%
Chemical Name or Material Cyclohexanecarbonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.