missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, Puriss. p.a., ACS Reagent, ≥99.5% (GC), Solstice

Product Code. 15663890
Change view
Click to view available options
Quantity:
1 L
2.5 L
5 L
30 kg
4 x 2.5 L
6 x 1 L
Packaging:
Aluminum Bottle
Glass Bottle
Stainless Steel Can
Unit Size:
1 L
2.5 L
30kg
5 L
Pack of 4
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Quantity Packaging Tamaño de la unidad
15663890 2.5 L Glass Bottle 2.5 L
15653890 1 L Glass Bottle 1 L
15613900 5 L Aluminum Bottle 5 L
15623900 6 x 1 L Glass Bottle Pack of 6
15693890 4 x 2.5 L Glass Bottle Pack of 4
15673890 30 kg Stainless Steel Can 30kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 15663890 Proveedor Honeywell-Riedel-de Haen N.º de proveedor 331172.5L
  • ONLINE EXCLUSIVE PRICE
    £102.00
    £80.40 / 2.5 L
    Save £21.60
    21% Off
 Restricted Item
In Stock
Añadir a la cesta
Añadir a la cesta
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Puriss. p.a., ACS Reagent,≥99.5% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Especificaciones

Melting Point 4°C to 7°C
Density 0.779g/mL at 25°C
Quantity 2.5 L
Explosion Limit 0.09
Flash Point −18°C (−.4°F)
Ignition Point 500°F
Packaging Glass Bottle
Refractive Index n20/D 1.4250 to 1.4280
Residue after Evaporation 0 to 0.001%
Vapor Pressure 168.8mmHg (37.7°C), 77mmHg (20°C)
UN Number UN1145
Beilstein 1900225
Boiling Point 79°C to 81°C
Formula Weight 84.16g/mol
Percent Purity ≥99.5% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material Cyclohexane
Mostrar más Mostrar menos
compliance-icons compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Cyclohexane
Palabra de advertencia
  • Danger
Clasificación
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Otros peligros
  • MIXTURE LIST-Contain: cyclohexane
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.