missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, HPLC Grade, 99% min

Product Code. 11358187
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
Unit Size:
1 L
4 L
This item is not returnable. View return policy

Product Code. 11358187

Brand: Thermo Scientific Alfa Aesar 022919.K7

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 110-82-7 | C6H12 | 84.16 g/mol

Cyclohexane is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Melting Point 6.5°C
Color Colorless
Appearance Colorless liquid
Density 0.779 g/cm3 at 20°C
Boiling Point 80°C to 81°C
Quantity 4 L
Flash Point -18°C (-1°F)
Odor Pungent
Physical Form Liquid
Refractive Index 1.426
Assay Percent Range 99% min.
Vapor Pressure 104 hPa (78mm Hg) at 20°C
Beilstein 1900225
Solubility Information Immiscible with water and soluble in ether,alcohol and acetone. Miscible with olive oil.
Formula Weight 84.16
Grade HPLC
Chemical Name or Material Cyclohexane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.