missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, HPLC Grade, 99% min, Thermo Scientific Chemicals

Catalog Number 11368187
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
110-82-7
C6H12
84.16
MFCD00003814
XDTMQSROBMDMFD-UHFFFAOYSA-N
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
8078
CHEBI:29005
cyclohexane
C1CCCCC1
110-82-7
C6H12
84.16
MFCD00003814
XDTMQSROBMDMFD-UHFFFAOYSA-N
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
8078
CHEBI:29005
cyclohexane
C1CCCCC1

Cyclohexane is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 6.5°C
Color Colorless
Appearance Colorless liquid
Density 0.779
Boiling Point 80°C to 81°C
Quantity 1 L
Flash Point -18°C (-1°F)
Odor Pungent
Physical Form Liquid
Refractive Index 1.426
Assay Percent Range 99% min.
Vapor Pressure 104 hPa (78mm Hg) at 20°C
Beilstein 1900225
Solubility Information Immiscible with water and soluble in ether,alcohol and acetone. Miscible with olive oil.
Formula Weight 84.16
Grade HPLC
Chemical Name or Material Cyclohexane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'