Learn More
Cyclohexane, Certified AR for Analysis, Fisher Chemical™
C6H12, CAS Number-110-82-7, 2.5L, 81 deg.C, CHEBI:29005, Colorless, 84.15g/mol, XDTMQSROBMDMFD-UHFFFAOYSA-N, cyclohexane, 6.5 deg.C, 3814, 84.162, Amber glass bottle, Liquid, 8078, C1CCCCC1, 2.9, 104mbar at 20 deg.C, 0.94 mPaS at 20 deg.C
Brand: Fisher Chemical C/8921/21
Code : 22
Additional Details : CAS Number : 110-82-7 Weight : 3.89500kg Transport : UN number : 1145 Chem class : 3 Pack group : II
Description
C6H12, CAS Number-110-82-7, 2.5L, 81 deg.C, CHEBI:29005, Colorless, 84.15g/mol, XDTMQSROBMDMFD-UHFFFAOYSA-N, cyclohexane, 6.5 deg.C, 3814, 84.162, Amber glass bottle, Liquid, 8078, C1CCCCC1, 2.9, 104mbar at 20 deg.C, 0.94 mPaS at 20 deg.C
Specifications
110-82-7 | |
3814 | |
Liquid | |
104mbar at 20°C | |
2.9 | |
81°C | |
5L |
C6H12 | |
84.15g/mol | |
Metal can | |
0.94 mPaS at 20°C | |
6.5°C | |
Colorless |
Safety and Handling
- Cyclohexane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.