missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, BAKER ULTRA RESI-ANALYZED, J.T. Baker™

Product Code. 10035161 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10035161 2.5 L Glass 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10035161 Supplier JT Baker Supplier No. 9258.2500
  • ONLINE EXCLUSIVE PRICE
    £403.00
    £288.73 / 2.5 L
    Save £114.27
    28% Off
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Gives superior performance with minimal risk of contaminants that can limit accuracy and productivity. Cyclohexane, BAKER ULTRA RESI-ANALYZED, J.T. Baker™ is an ultra-pure solvent for organic residue analysis that maximizes an instrument’s sensitivity and detection power.

BAKER ULTRA RESI-ANALYZED solvents were developed for organic residue extraction/concentration procedures. These solvents are fully characterized and lot controlled by ECD, FID, or other method-specific detectors to deliver the highest level of purity and lot-to-lot consistency.

  • Unique stabilizer systems provide unmatched product stability and interference-free results when chemical requires
  • Optimized distillation targeted to remove reactive impurities, production of higher assays and narrow solvent fronts
  • Performance tested to purity levels below the Lower Limits of Quantitation (LLQ) for trace analyte detection
  • Performance is guaranteed through the expiration period stated on the label
  • Suitable for exacting EPA sample cleanup protocols

Specifications

Melting Point 6.4°C
CAS 110-82-7
Color Colorless
Density 0.78g/cm3 at 20°C
Quantity 2.5 L
Explosion Limit 1.2 - 8.3 Vol %
Flash Point −18°C
Ignition Point 260°C
Odor Sweetish
Packaging Glass
Physical Form Liquid
Residue after Evaporation Max. 2 ppm
Molecular Formula C6H12
Linear Formula CH2(CH2)4CH2
MDL Number MFCD00003814
Viscosity 0.94 mPas at 20°C
Water Max. 100 ppm
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
Boiling Point 81°C
Solubility Information 0.05g/L (in water)
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
SMILES C1CCCCC1
IUPAC Name cyclohexane
Molecular Weight (g/mol) 84.16
PubChem CID 8078
ChEBI CHEBI:29005
Formula Weight 84.16g/mol
Percent Purity ≥99.0%
Grade For Organic Residue Analysis
Assay Corrected for water
Chemical Name or Material Cyclohexane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.